Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nitr12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025496.1 | NP_001020667.1 | 3 - 4 | 767 | 78 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |