Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tspan3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001025538.2 | NP_001020709.1 | 5 - 6 | 694 | 86 | |
GenBank mRNA | BC097114.1 | - | 5 - 6 | 691 | 86 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |