Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lgi2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039641.1 | NP_001034730.1 | 5 - 6 | 521 | 104 | |
GenBank mRNA | AY309262.1 | - | 5 - 6 | 521 | 104 | |
BC162305.1 | - | 5 - 6 | 493 | 104 | ||
BC162306.1 | - | 5 - 6 | 493 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Danio rerio Probe spans exons |