Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tnfsf10l3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001042713.1 | NP_001036178.1 | 1 - 2 | 133 | 69 | |
GenBank mRNA | BC162834.1 | - | 1 - 2 | 215 | 69 | |
BC162841.1 | - | 1 - 2 | 215 | 69 | ||
DQ812113.1 | - | 1 - 2 | 133 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Inventoried Probe spans exons |