Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044313.2 | NP_001037778.2 | 5 - 6 | 417 | 129 | |
XM_005173946.4 | XP_005174003.1 | 6 - 7 | 813 | 129 | ||
XM_009296096.3 | XP_009294371.1 | 6 - 7 | 813 | 129 | ||
GenBank mRNA | AW174241.1 | - | 5 - 6 | 409 | 129 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Danio rerio Probe spans exons |