Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tmcc3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044813.2 | NP_001038278.1 | 1 - 2 | 227 | 73 | |
GenBank mRNA | BC133824.1 | - | 1 - 2 | 227 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |