Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: furinb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045109.2 | NP_001038574.2 | 5 - 6 | 1059 | 77 | |
GenBank mRNA | BC163243.1 | - | 5 - 6 | 769 | 77 | |
DQ499477.1 | - | 5 - 6 | 1019 | 77 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |