Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snap47 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045437.1 | NP_001038902.1 | 3 - 4 | 1186 | 62 | |
XM_021481304.1 | XP_021336979.1 | 3 - 4 | 1346 | 62 | ||
GenBank mRNA | BC122195.1 | - | 3 - 4 | 1186 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |