Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: alkbh5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001077387.1 | NP_001070855.1 | 3 - 4 | 1244 | 93 | |
GenBank mRNA | BC124804.1 | - | 3 - 4 | 1244 | 93 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |