Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329567.1 | NP_001316496.1 | 3 - 4 | 280 | 62 | |
XM_021478298.1 | XP_021333973.1 | 3 - 4 | 336 | 62 | ||
GenBank mRNA | BC154484.1 | - | 3 - 4 | 280 | 62 | |
BC163356.1 | - | 2 - 3 | 259 | 62 | ||
FJ906821.1 | - | 2 - 3 | 186 | 62 | ||
FM881778.1 | - | 2 - 3 | 253 | 62 | ||
FM881779.1 | - | 2 - 3 | 253 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |