Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: foxp3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001329567.1 | NP_001316496.1 | 8 - 9 | 796 | 83 | |
XM_021478298.1 | XP_021333973.1 | 8 - 9 | 852 | 83 | ||
GenBank mRNA | BC154484.1 | - | 8 - 9 | 796 | 83 | |
BC163356.1 | - | 7 - 8 | 775 | 83 | ||
FM881778.1 | - | 7 - 8 | 769 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |