Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: flr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104649.2 | NP_001098119.2 | 14 - 15 | 1528 | 132 | |
GenBank mRNA | EU124003.1 | - | 14 - 15 | 1530 | 132 |
Set Membership: |
Amplicon length greater than or equal to 101 Danio rerio Probe spans exons |