Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cables1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105665.1 | NP_001099135.1 | 1 - 2 | 583 | 72 | |
XM_005163651.4 | XP_005163708.1 | 1 - 2 | 1108 | 72 | ||
XM_021478424.1 | XP_021334099.1 | 1 - 2 | 1108 | 72 | ||
GenBank mRNA | EF105292.1 | - | 1 - 2 | 583 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Danio rerio Probe spans exons |