Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cry2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_131786.2 | NP_571861.2 | 4 - 5 | 794 | 127 | |
GenBank mRNA | AB042252.1 | - | 4 - 5 | 683 | 127 | |
BC046088.1 | - | 4 - 5 | 794 | 127 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |