Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182861.1 | NP_878281.1 | 1 - 2 | 85 | 59 | |
GenBank mRNA | AY167023.1 | - | 1 - 2 | 85 | 59 | |
BC050950.1 | - | 1 - 2 | 61 | 59 | ||
BC163864.1 | - | 1 - 2 | 58 | 59 | ||
BC163870.1 | - | 1 - 2 | 58 | 59 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |