Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: scfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_182861.1 | NP_878281.1 | 9 - 10 | 776 | 74 | |
GenBank mRNA | AY167023.1 | - | 9 - 10 | 776 | 74 | |
BC050950.1 | - | 9 - 10 | 752 | 74 | ||
BC163864.1 | - | 9 - 10 | 749 | 74 | ||
BC163870.1 | - | 9 - 10 | 749 | 74 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |