Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: znhit3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200273.1 | NP_956567.1 | 4 - 5 | 334 | 71 | |
XM_021476032.1 | XP_021331707.1 | 4 - 5 | 358 | 71 | ||
GenBank mRNA | BC049063.1 | - | 4 - 5 | 334 | 71 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |