Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gorab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200385.1 | NP_956679.1 | 1 - 2 | 186 | 63 | |
XM_005160649.4 | XP_005160706.1 | 1 - 2 | 232 | 63 | ||
GenBank mRNA | BC053289.1 | - | 1 - 2 | 186 | 63 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |