Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gorasp2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200703.2 | NP_956997.1 | 9 - 10 | 1173 | 64 | |
XM_021476811.1 | XP_021332486.1 | 9 - 10 | 1158 | 64 | ||
GenBank mRNA | AY398354.1 | - | 9 - 10 | 1173 | 64 | |
BC059447.1 | - | 9 - 10 | 1134 | 64 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |