Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ppm1g | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201488.1 | NP_958896.1 | 5 - 6 | 664 | 91 | |
XM_005169512.4 | XP_005169569.1 | 4 - 5 | 832 | 91 | ||
GenBank mRNA | BC052132.1 | - | 5 - 6 | 664 | 91 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |