Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: epdr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002416.2 | NP_001002416.1 | 2 - 3 | 539 | 79 | |
GenBank mRNA | BC076158.1 | - | 2 - 3 | 541 | 79 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |