Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mfsd2ab | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001003570.2 | NP_001003570.1 | 5 - 6 | 750 | 71 | |
GenBank mRNA | BC077117.1 | - | 5 - 6 | 697 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |