Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dtwd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005951.2 | NP_001005951.2 | 3 - 4 | 623 | 87 | |
GenBank mRNA | BC083451.1 | - | 3 - 4 | 619 | 87 | |
EH463350.1 | - | 1 - 2 | 373 | 87 | ||
EH540548.1 | - | 3 - 4 | 649 | 87 | ||
EH553437.1 | - | 3 - 4 | 602 | 87 |
Gene Symbol: nnt2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017351991.2 | XP_017207480.1 | 3778 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Danio rerio Probe spans exons |