Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wacb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001005971.1 | NP_001005971.1 | 4 - 5 | 650 | 137 | |
GenBank mRNA | BC083294.1 | - | 4 - 5 | 650 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |