Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mcamb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001012367.2 | NP_001012367.2 | 8 - 9 | 1107 | 125 | |
XM_005157570.2 | XP_005157627.1 | 8 - 9 | 1130 | 125 | ||
XM_021481353.1 | XP_021337028.1 | 8 - 9 | 1127 | 125 | ||
GenBank mRNA | AY497061.1 | - | 8 - 9 | 1086 | 125 | |
AY746594.1 | - | 8 - 9 | 1107 | 125 | ||
BC162589.1 | - | 8 - 9 | 1099 | 125 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |