Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyp2x6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013565.2 | NP_001013583.1 | 7 - 8 | 1185 | 62 | |
GenBank mRNA | BC078308.1 | - | 7 - 8 | 1190 | 62 | |
BC090406.1 | - | 7 - 8 | 1189 | 62 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |