Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: acsf2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001045419.2 | NP_001038884.2 | 5 - 6 | 691 | 83 | |
NM_001139438.1 | NP_001132910.1 | 4 - 5 | 851 | 83 | ||
GenBank mRNA | BC122098.1 | - | 5 - 6 | 686 | 83 | |
CF348078.1 | - | 1 - 2 | 73 | 83 | ||
EH554699.1 | - | 4 - 5 | 892 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |