Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: btr04 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_017356979.2 | XP_017212468.1 | 6 - 7 | 1058 | 85 | |
GenBank mRNA | BC122135.1 | - | 6 - 7 | 948 | 85 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Probe spans exons |