Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mixl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | BC097091.1 | - | 3 - 4 | 1470 | 139 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |