Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ucp3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200353.2 | NP_956647.2 | 6 - 7 | 934 | 78 | |
GenBank mRNA | BC053173.1 | - | 6 - 7 | 904 | 77 | |
BC154331.1 | - | 6 - 7 | 897 | 78 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |