Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: akt2l | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212815.2 | NP_997980.1 | 11 - 12 | 1346 | 71 | |
XM_021479143.1 | XP_021334818.1 | 1429 | 71 | |||
XM_021479144.1 | XP_021334819.1 | 1437 | 71 | |||
XM_021479145.1 | XP_021334820.1 | 681 | 71 | |||
XM_021479146.1 | XP_021334821.1 | 363 | 71 | |||
GenBank mRNA | BC046892.1 | - | 11 - 12 | 1347 | 71 | |
BC068106.1 | - | 11 - 12 | 1314 | 71 | ||
BC165215.1 | - | 10 - 11 | 1196 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |