Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sp9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_212960.2 | NP_998125.2 | 1 | 1749 | 137 | |
XM_005167414.4 | XP_005167471.1 | 2 | 1311 | 137 | ||
GenBank mRNA | AY591905.1 | - | 1 | 1749 | 137 | |
AY731230.1 | - | 2 | 1265 | 137 | ||
BC067673.1 | - | 2 | 1195 | 137 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Mammalian Gene Collection Detect Genomic DNA Recommended Assay Danio rerio Both primers and probe map within a single exon |