Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: syncrip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214696.1 | NP_999861.1 | 3 - 4 | 387 | 69 | |
XM_009294525.1 | XP_009292800.1 | 3 - 4 | 451 | 69 | ||
XM_009294526.1 | XP_009292801.1 | 3 - 4 | 451 | 69 | ||
XM_009294527.1 | XP_009292802.1 | 2 - 3 | 280 | 69 | ||
GenBank mRNA | BC056750.1 | - | 3 - 4 | 387 | 69 | |
BC068373.1 | - | 3 - 4 | 384 | 69 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |