Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pcnp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006035.2 | NP_001006035.1 | 3 - 4 | 441 | 116 | |
NM_001326716.1 | NP_001313645.1 | 4 - 5 | 400 | 116 | ||
NM_001326717.1 | NP_001313646.1 | 3 - 4 | 373 | 116 | ||
NM_001326718.1 | NP_001313647.1 | 3 - 4 | 370 | 116 | ||
NM_001326719.1 | NP_001313648.1 | 4 - 5 | 396 | 116 | ||
XM_021473273.1 | XP_021328948.1 | 4 - 5 | 399 | 116 | ||
GenBank mRNA | BC083380.1 | - | 3 - 4 | 419 | 116 |
Gene Symbol: LOC110437793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021471049.1 | XP_021326724.1 | 2 - 3 | 289 | 116 | |
XM_021471050.1 | XP_021326725.1 | 3 - 4 | 698 | 116 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |