Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: golph3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001006105.1 | NP_001006105.1 | 1 - 2 | 491 | 68 | |
GenBank mRNA | BC083206.1 | - | 1 - 2 | 491 | 68 | |
BC165929.1 | - | 1 - 2 | 250 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Danio rerio Probe spans exons |