Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fgf18a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001013264.1 | NP_001013282.1 | 3 - 4 | 253 | 60 | |
XM_021480985.1 | XP_021336660.1 | 3 - 4 | 1840 | 60 | ||
GenBank mRNA | AB195467.1 | - | 3 - 4 | 253 | 60 | |
AY243514.1 | - | 1 - 2 | 184 | 60 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Recommended Assay Danio rerio Probe spans exons |