Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: bmpr2b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001039807.1 | NP_001034896.1 | 5 - 6 | 1432 | 95 | |
GenBank mRNA | BC163200.1 | - | 5 - 6 | 95 | ||
DQ002899.1 | - | 5 - 6 | 1432 | 95 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |