Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ptpn23b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001044928.1 | NP_001038393.1 | 15 - 16 | 1536 | 109 | |
XM_005159517.4 | XP_005159574.1 | 15 - 16 | 1525 | 109 | ||
GenBank mRNA | BC171436.1 | - | 15 - 16 | 1446 | 109 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |