Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hcrtr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001079868.1 | NP_001073337.1 | 4 - 5 | 765 | 101 | |
GenBank mRNA | EF122429.1 | - | 4 - 5 | 765 | 101 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Danio rerio Probe spans exons |