Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubfd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001082830.1 | NP_001076299.1 | 3 - 4 | 519 | 87 | |
XM_005164210.4 | XP_005164267.1 | 3 - 4 | 603 | 87 | ||
GenBank mRNA | BC134031.1 | - | 3 - 4 | 519 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |