Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lctlb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001100014.2 | NP_001093484.1 | 6 - 7 | 689 | 96 | |
XM_005159111.3 | XP_005159168.1 | 7 - 8 | 908 | 96 | ||
XM_009293569.2 | XP_009291844.1 | 7 - 8 | 908 | 96 | ||
XM_021467671.1 | XP_021323346.1 | 7 - 8 | 908 | 96 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Danio rerio Probe spans exons |