Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nos2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001104937.1 | NP_001098407.1 | 9 - 10 | 1104 | 66 | |
XM_005165296.4 | XP_005165353.1 | 9 - 10 | 1172 | 66 | ||
GenBank mRNA | AM084880.1 | - | 9 - 10 | 1104 | 66 | |
AM749801.1 | - | 9 - 10 | 1104 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Danio rerio Probe spans exons |