Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: chrd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_130973.3 | NP_571048.2 | 14 - 15 | 2302 | 73 | |
GenBank mRNA | AF034606.1 | - | 14 - 15 | 2304 | 73 | |
BC162594.1 | - | 14 - 15 | 2304 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Mammalian Gene Collection Danio rerio Probe spans exons |