Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rmnd5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_200774.1 | NP_957068.1 | 5 - 6 | 577 | 103 | |
GenBank mRNA | BC059606.1 | - | 5 - 6 | 577 | 103 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |