Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubtfl | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201003.1 | NP_957297.1 | 16 - 17 | 1876 | 100 | |
XM_005163486.3 | XP_005163543.1 | 16 - 17 | 2801 | 100 | ||
XM_005163487.3 | XP_005163544.1 | 16 - 17 | 2786 | 100 | ||
XM_005163488.3 | XP_005163545.1 | 16 - 17 | 2783 | 100 | ||
XM_005163489.3 | XP_005163546.1 | 16 - 17 | 2801 | 100 | ||
XM_005163490.3 | XP_005163547.1 | 16 - 17 | 2783 | 100 | ||
XM_009298612.2 | XP_009296887.1 | 16 - 17 | 2798 | 100 | ||
XM_009298613.2 | XP_009296888.1 | 16 - 17 | 2801 | 100 | ||
XM_009298614.2 | XP_009296889.1 | 16 - 17 | 2801 | 100 | ||
XM_017357972.2 | XP_017213461.1 | 15 - 16 | 1827 | 100 | ||
GenBank mRNA | BC055140.1 | - | 16 - 17 | 1876 | 100 |
Set Membership: |
Amplicon length between 86 and 100 Mammalian Gene Collection Danio rerio Probe spans exons |