Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: snrpe | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201004.1 | NP_957298.1 | 4 - 5 | 351 | 84 | |
GenBank mRNA | BC049062.1 | - | 4 - 5 | 351 | 84 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |