Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201468.1 | NP_958876.1 | 4 - 5 | 599 | 142 | |
XM_005161091.4 | XP_005161148.1 | 4 - 5 | 617 | 142 | ||
GenBank mRNA | AY398416.1 | - | 4 - 5 | 599 | 142 | |
BC066410.1 | - | 4 - 5 | 553 | 142 |
Set Membership: |
Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Danio rerio Probe spans exons |