Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: setb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_201468.1 | NP_958876.1 | 6 - 7 | 874 | 185 | |
GenBank mRNA | AY398416.1 | - | 6 - 7 | 874 | 185 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Danio rerio Probe spans exons |