Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: sdr16c5b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_214771.2 | NP_999936.1 | 6 - 7 | 967 | 65 | |
GenBank mRNA | BC054138.1 | - | 6 - 7 | 969 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |