Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: faah2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001002700.2 | NP_001002700.1 | 1 - 2 | 251 | 65 | |
GenBank mRNA | BC076113.1 | - | 1 - 2 | 252 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Danio rerio Probe spans exons |